Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUFY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040451.2 | NP_001035541.1 | 17 | 2519 | ||
NM_001040452.2 | NP_001035542.1 | 18 | 2330 | |||
NM_025158.4 | NP_079434.3 | 18 | 2505 | |||
XM_005265993.4 | XP_005266050.1 | 15 | 1876 | |||
XM_006714921.3 | XP_006714984.2 | 17 | 2449 | |||
XM_006714922.3 | XP_006714985.1 | 15 | 2309 | |||
XM_017009891.1 | XP_016865380.1 | 17 | 2264 | |||
XM_017009892.1 | XP_016865381.1 | 16 | 2302 | |||
XM_017009893.1 | XP_016865382.1 | 15 | 2236 | |||
XM_017009894.1 | XP_016865383.1 | 19 | 2471 | |||
XM_017009895.1 | XP_016865384.1 | 18 | 2455 | |||
GenBank mRNA | AF312367.1 | - | 18 | 2496 | ||
AF361055.2 | - | 18 | 2478 | |||
AK025904.1 | - | 12 | 1568 | |||
AK075021.1 | - | 17 | 2418 | |||
AK093848.1 | - | 10 | 2437 | |||
BC032571.1 | - | 18 | 2249 | 5243223 |