Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35F5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025181.3 | NP_079457.2 | 7 | 1034 | ||
NR_104470.1 | 7 | 1419 | ||||
XM_005263799.3 | XP_005263856.1 | 7 | 1419 | |||
XM_011511921.2 | XP_011510223.1 | 7 | 831 | |||
XM_011511922.2 | XP_011510224.1 | 7 | 1048 | |||
XM_011511923.2 | XP_011510225.1 | 7 | 831 | |||
XM_017005027.1 | XP_016860516.1 | 7 | 831 | |||
XM_017005028.1 | XP_016860517.1 | 7 | 831 | |||
GenBank mRNA | AF529364.1 | - | 7 | 620 | ||
AK024666.1 | - | 2 | 161 | |||
AK025657.1 | - | 7 | 1346 | |||
AK315719.1 | - | 7 | 832 | |||
AY358620.1 | - | 7 | 777 | |||
BC018537.1 | - | 7 | 1044 | 4340152 |