Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272004.1 | NP_001258933.1 | 5 | 1091 | ||
NM_001272019.2 | NP_001258948.1 | 5 | 697 | |||
NM_001282391.1 | NP_001269320.1 | 5 | 943 | |||
NM_025209.3 | NP_079485.1 | 5 | 1091 | |||
XM_006717510.1 | XP_006717573.1 | 5 | 943 | |||
GenBank mRNA | AF277374.1 | - | 5 | 1058 | ||
AF286905.1 | - | 5 | 761 | |||
AK024329.1 | - | 2 | 206 | |||
AK092304.1 | - | 5 | 697 | |||
AK299676.1 | - | 5 | 884 | |||
AK309309.1 | - | 5 | 829 | |||
AK309822.1 | - | 5 | 960 | |||
BC036529.1 | - | 5 | 1092 | 4828057 | ||
CR749375.1 | - | 6 | 785 |