Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other STK33 SIRNA Assays ›
Gene Symbol:
STK33
Gene Name:
serine/threonine kinase 33
Gene Aliases:
-
Chromosome Location:
Chr.11: 8335029 - 8594289 on Build GRCh38
UniGene ID:
Hs.501833
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
131006
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: STK33
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001289058.1
NP_001275987.1
4
405
NM_001289061.1
NP_001275990.1
3
670
NM_030906.3
NP_112168.1
4
819
XM_011520288.2
XP_011518590.1
4
1897
XM_011520290.2
XP_011518592.1
6
816
XM_011520292.2
XP_011518594.1
4
618
XM_011520293.2
XP_011518595.1
4
647
XM_011520294.1
XP_011518596.1
3
612
XM_011520295.2
XP_011518597.1
3
2708
XM_011520297.2
XP_011518599.1
3
385
XM_017018143.1
XP_016873632.1
6
876
XM_017018144.1
XP_016873633.1
6
778
XM_017018145.1
XP_016873634.1
5
2810
XM_017018146.1
XP_016873635.1
6
940
XM_017018147.1
XP_016873636.1
5
2737
XM_017018148.1
XP_016873637.1
4
2669
XM_017018149.1
XP_016873638.1
5
821
XM_017018150.1
XP_016873639.1
5
676
XM_017018151.1
XP_016873640.1
5
635
XM_017018152.1
XP_016873641.1
3
966
XM_017018153.1
XP_016873642.1
3
564
XM_017018154.1
XP_016873643.1
3
423
XM_017018155.1
XP_016873644.1
3
978
XM_017018156.1
XP_016873645.1
3
424
XM_017018157.1
XP_016873646.1
3
189
XM_017018158.1
XP_016873647.1
4
475
XM_017018159.1
XP_016873648.1
5
2813
XM_017018160.1
XP_016873649.1
3
612
XM_017018161.1
XP_016873650.1
5
2819
XM_017018162.1
XP_016873651.1
4
422
XM_017018163.1
XP_016873652.1
3
413
XM_017018164.1
XP_016873653.1
3
299
XM_017018165.1
XP_016873654.1
4
378
GenBank mRNA
AJ303380.2
-
2
1163
AK093251.1
-
3
528
AK293193.1
-
4
271
BC031231.1
-
4
767
5296502
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein autophosphorylation
protein serine/threonine kinase activity
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-2g8tf:80/10.239.165.190:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0