Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6orf62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030939.4 | NP_112201.1 | 5 | 1175 | ||
XM_011514928.2 | XP_011513230.1 | 5 | 1516 | |||
GenBank mRNA | AB075502.1 | - | 5 | 1162 | ||
AJ420475.1 | - | 3 | 354 | |||
AK022681.1 | - | 5 | 1059 | |||
AK301718.1 | - | 5 | 714 | |||
AL136632.2 | - | 5 | 1186 | |||
AY598792.1 | - | 5 | 668 | |||
BC040260.1 | - | 5 | 687 | |||
BC047866.1 | - | 5 | 866 | 6499979 | ||
CR457292.1 | - | 5 | 668 | |||
CR533486.1 | - | 5 | 668 |