Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLVAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031310.2 | NP_112600.1 | 3 | 599 | ||
GenBank mRNA | AF326591.1 | - | 3 | 590 | ||
AF348827.1 | - | 3 | 588 | |||
AK096030.1 | - | 3 | 592 | |||
AK313837.1 | - | 3 | 592 | |||
AL834363.1 | - | 3 | 581 | |||
BC050365.1 | - | 3 | 578 | 5761483 | ||
BC056414.1 | - | 3 | 600 | 6190596 | ||
BX380375.2 | - | 3 | 600 |