Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLVAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031310.2 | NP_112600.1 | 6 | 1687 | ||
GenBank mRNA | AF326591.1 | - | 6 | 1678 | ||
AF348827.1 | - | 6 | 1676 | |||
AK074054.1 | - | 4 | 769 | |||
AK096030.1 | - | 6 | 1665 | |||
AL834363.1 | - | 6 | 1669 | |||
BC050365.1 | - | 6 | 1666 | 5761483 | ||
BC056414.1 | - | 6 | 1688 | 6190596 | ||
D83199.1 | - | 1 | 147 |