Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SESN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031459.4 | NP_113647.1 | 8 | 1512 | ||
GenBank mRNA | AK025640.1 | - | 2 | 137 | ||
AK027896.1 | - | 6 | 825 | |||
AK315710.1 | - | 8 | 1225 | |||
AL136551.1 | - | 8 | 1292 | |||
AY123223.1 | - | 8 | 1438 | |||
BC013304.2 | - | 8 | 1440 | 2959845 | ||
BC033719.1 | - | 8 | 1446 | 4931535 |