Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131028.1 | NP_001124500.1 | 8 | 1018 | ||
NM_031482.4 | NP_113670.1 | 7 | 883 | |||
XM_005248610.4 | XP_005248667.1 | 7 | 2153 | |||
XM_005248611.4 | XP_005248668.1 | 7 | 727 | |||
XM_005248612.3 | XP_005248669.1 | 6 | 779 | |||
XM_011543660.2 | XP_011541962.1 | 6 | 583 | |||
XM_011543661.2 | XP_011541963.1 | 5 | 966 | |||
XM_017009944.1 | XP_016865433.1 | 7 | 914 | |||
GenBank mRNA | AK225969.1 | - | 6 | 775 | ||
AK315752.1 | - | 7 | 883 | |||
AL136912.1 | - | 7 | 751 | |||
BC029268.1 | - | 7 | 853 | 4521833 |