Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037290.2 | NP_001032367.2 | 3 | 427 | ||
NM_001256397.1 | NP_001243326.1 | 3 | 427 | |||
NM_001256398.1 | NP_001243327.1 | 3 | 539 | |||
NM_001256400.1 | NP_001243329.1 | 2 | 222 | |||
NM_031938.5 | NP_114144.4 | 3 | 539 | |||
GenBank mRNA | AB096252.1 | - | 3 | 393 | ||
AF276432.1 | - | 3 | 1036 | |||
AJ290393.1 | - | 3 | 389 | |||
AK027801.1 | - | 3 | 422 | |||
AK091783.1 | - | 2 | 274 | |||
AK297853.1 | - | 2 | 213 | |||
AK315300.1 | - | 3 | 531 | |||
BC041656.1 | - | 3 | 502 | 5175925 | ||
BX503569.1 | - | 3 | 427 |