Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032271.2 | NP_115647.2 | 11, 12 | 1178 | ||
XM_005255627.4 | XP_005255684.1 | 11, 12 | 1218 | |||
XM_011522700.1 | XP_011521002.1 | 11, 12 | 1220 | |||
GenBank mRNA | AK090624.1 | - | 11, 12 | 1192 | ||
AL136921.1 | - | 10, 11 | 1074 | |||
AY569455.1 | - | 10, 11 | 1131 | |||
BC024267.1 | - | 9, 10 | 964 |