Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278279.2 | NP_001265208.2 | 4 | 611 | ||
NM_032351.5 | NP_115727.5 | 4 | 611 | |||
GenBank mRNA | AK172741.1 | - | 4 | 471 | ||
AK225031.1 | - | 4 | 471 | |||
AK293605.1 | - | 4 | 577 | |||
BC130382.1 | - | 4 | 502 | 40146413 | ||
BC130384.1 | - | 4 | 502 | 40146415 | ||
BF240534.1 | - | 607 | ||||
BG719833.1 | - | 4 | 488 | |||
BG761720.1 | - | 4 | 471 | |||
BI767984.1 | - | 4 | 452 |
Gene Symbol: MRPL45P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033934.1 | 4 | 623 | |||
GenBank mRNA | BC090855.1 | - | 5 | 593 |