Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCGF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256549.1 | NP_001243478.1 | 8, 9 | 1023 | ||
NM_001257101.1 | NP_001244030.1 | 8, 9 | 916 | |||
NM_032373.4 | NP_115749.2 | 8, 9 | 1081 | |||
XM_006718030.1 | XP_006718093.1 | 7, 8 | 1012 | |||
XM_011540271.2 | XP_011538573.1 | 8, 9 | 1082 | |||
XM_017016776.1 | XP_016872265.1 | 8, 9 | 1028 | |||
GenBank mRNA | AK303014.1 | - | 8, 9 | 1024 | ||
AL832003.2 | - | 6, 7 | 520 | |||
AL832496.1 | - | 8, 9 | 897 | |||
BC051845.1 | - | 8, 9 | 891 | 5199133 |