Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258452.1 | NP_001245381.1 | 10 | 1834 | ||
NM_001258453.1 | NP_001245382.1 | 11 | 1906 | |||
NM_032807.4 | NP_116196.3 | 11 | 1922 | |||
NM_178150.2 | NP_835363.1 | 10 | 1808 | |||
XM_011519748.2 | XP_011518050.1 | 12 | 2502 | |||
XM_017016842.1 | XP_016872331.1 | 11 | 3049 | |||
XM_017016843.1 | XP_016872332.1 | 12 | 2433 | |||
XM_017016844.1 | XP_016872333.1 | 10 | 1818 | |||
XM_017016845.1 | XP_016872334.1 | 10 | 2175 | |||
XM_017016846.1 | XP_016872335.1 | 9 | 1627 | |||
GenBank mRNA | AF380349.1 | - | 11 | 1922 | ||
AF454502.1 | - | 10 | 1749 | |||
AF456237.1 | - | 10 | 1764 | |||
AK027496.1 | - | 7 | 891 | |||
AK095343.1 | - | 10 | 1782 | |||
AK122753.1 | - | 10 | 1834 | |||
AK226128.1 | - | 10 | 2332 | |||
AK315173.1 | - | 9 | 1524 | |||
BC020266.1 | - | 8 | 1316 | |||
BC032674.1 | - | 8 | 1260 | |||
BC080629.1 | - | 8 | 1471 | |||
BC110884.1 | - | 11 | 1831 | 6147373 | ||
BC113375.1 | - | 10 | 1755 | 8322427 | ||
BC113377.1 | - | 10 | 1755 | 8322429 |