Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DISP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032890.3 | NP_116279.2 | 4 | 547 | ||
XM_005273335.2 | XP_005273392.1 | 2 | 388 | |||
XM_006711592.2 | XP_006711655.1 | 3 | 1335 | |||
XM_011510072.2 | XP_011508374.1 | 3 | 387 | |||
XM_011510073.2 | XP_011508375.1 | 3 | 865 | |||
XM_011510074.2 | XP_011508376.1 | 3 | 406 | |||
XM_011510075.2 | XP_011508377.1 | 3 | 893 | |||
XM_011510077.2 | XP_011508379.1 | 2 | 329 | |||
XM_017002611.1 | XP_016858100.1 | 2 | 803 | |||
XM_017002613.1 | XP_016858102.1 | 1 | 14 | |||
GenBank mRNA | AK098669.1 | - | 2 | 281 | ||
AK125728.1 | - | 2 | 385 | |||
BC007734.2 | - | 2 | 368 | |||
BC107762.1 | - | 2 | 372 |