Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATCAY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033064.4 | NP_149053.1 | 5 | 930 | ||
GenBank mRNA | AB058775.1 | - | 5 | 876 | ||
AK027889.1 | - | 5 | 864 | |||
AK092309.1 | - | 5 | 729 | |||
AK125457.1 | - | 5 | 451 | |||
AK299522.1 | - | 6 | 936 | |||
AY220297.1 | - | 4 | 500 | |||
BC026217.1 | - | 5 | 872 | 4153341 | ||
KU178814.1 | - | 4 | 500 | |||
KU178815.1 | - | 4 | 500 |