Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144766.2 | NP_001138238.1 | 11 | 1193 | ||
NM_003315.3 | NP_003306.3 | 11 | 1425 | |||
NR_029431.1 | 10 | 1336 | ||||
XM_011525167.2 | XP_011523469.1 | 12 | 1355 | |||
XM_011525168.2 | XP_011523470.1 | 12 | 1315 | |||
XM_011525169.2 | XP_011523471.1 | 11 | 1167 | |||
XM_017024994.1 | XP_016880483.1 | 12 | 1349 | |||
GenBank mRNA | AB209834.1 | - | 11 | 1111 | ||
AK291047.1 | - | 11 | 1126 | |||
AK296602.1 | - | 11 | 1186 | |||
AK298860.1 | - | 11 | 1141 | |||
AK316081.1 | - | 10 | 1062 | |||
BC003601.1 | - | 10 | 1048 | 3543074 | ||
BC011837.2 | - | 11 | 1144 | 3945721 | ||
BC033772.1 | - | 11 | 1170 | 3924081 | ||
BX647209.1 | - | 11 | 1381 | |||
U46571.1 | - | 11 | 1140 |