Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGME1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310338.1 | NP_001297267.1 | 6 | 1402 | ||
NM_001310339.1 | NP_001297268.1 | 3 | 1004 | |||
NM_052865.3 | NP_443097.1 | 5 | 1357 | |||
XM_005260867.2 | XP_005260924.1 | 6 | 1624 | |||
XM_005260870.3 | XP_005260927.1 | 4 | 1172 | |||
XM_006723663.3 | XP_006723726.1 | 5 | 1217 | |||
XM_011529394.1 | XP_011527696.1 | 6 | 1239 | |||
XM_011529395.1 | XP_011527697.1 | 5 | 1117 | |||
XM_017028127.1 | XP_016883616.1 | 5 | 1579 | |||
XM_017028128.1 | XP_016883617.1 | 5 | 1453 | |||
GenBank mRNA | AF306685.1 | - | 5 | 1166 | ||
AK027503.1 | - | 5 | 1161 | |||
AK309563.1 | - | 3 | 700 | |||
AK309995.1 | - | 3 | 628 | |||
AK310282.1 | - | 4 | 673 | |||
BC016869.1 | - | 5 | 1129 | 3850853 |