Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIST1H2AH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080596.2 | NP_542163.1 | 1 | 443 | ||
GenBank mRNA | BC093849.1 | - | 1 | 443 | 7939694 | |
BC093851.1 | - | 1 | 443 | 7939696 | ||
BC134366.1 | - | 1 | 396 | 40021708 | ||
BC148293.2 | - | 1 | 396 | 40021709 |