Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSWIM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080603.4 | NP_542170.3 | 2 | 1567 | ||
XM_005260610.4 | XP_005260667.1 | 2 | 1701 | |||
XM_005260611.4 | XP_005260668.1 | 2 | 1784 | |||
XM_011529100.2 | XP_011527402.1 | 2 | 1658 | |||
GenBank mRNA | AK291304.1 | - | 2 | 1555 | ||
BC001672.2 | - | 2 | 1535 | 2988536 | ||
BC080525.1 | - | 2 | 1535 | 4547006 |