Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOZ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122853.2 | NP_001116325.1 | 6 | 1163 | ||
NM_133371.4 | NP_588612.2 | 6 | 815 | |||
GenBank mRNA | AF480443.1 | - | 6 | 667 | ||
AJ300586.1 | - | 6 | 778 | |||
AK313873.1 | - | 6 | 819 | |||
BC069380.1 | - | 6 | 645 | 7262195 | ||
BC069553.1 | - | 6 | 645 | 7262207 | ||
BC074870.2 | - | 6 | 601 | 30915232 | ||
BC074871.2 | - | 6 | 601 | 30915469 | ||
BP360917.1 | - | 2 | 299 |