Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270679.1 | NP_001257608.1 | 7 | 846 | ||
NM_001270680.1 | NP_001257609.1 | 7 | 1186 | |||
NM_138357.2 | NP_612366.1 | 7 | 909 | |||
NR_073062.1 | 9 | 1074 | ||||
XM_017016882.1 | XP_016872371.1 | 7 | 1109 | |||
GenBank mRNA | AK025699.1 | - | 3 | 251 | ||
AK075439.1 | - | 7 | 896 | |||
AK128016.1 | - | 7 | 1186 | |||
AK301475.1 | - | 9 | 1055 | |||
AK315519.1 | - | 7 | 892 | |||
BC010682.1 | - | 7 | 803 | |||
BC034235.1 | - | 7 | 909 | 5296557 |