Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173512.1 | NP_001166983.1 | 3 | 591 | ||
NM_138777.3 | NP_620132.1 | 4 | 747 | |||
NM_199177.2 | NP_954646.1 | 3 | 411 | |||
XM_005252302.3 | XP_005252359.1 | 5 | 815 | |||
XM_011519183.2 | XP_011517485.1 | 5 | 1070 | |||
XM_011519184.2 | XP_011517486.1 | 5 | 580 | |||
XM_011519185.2 | XP_011517487.1 | 5 | 815 | |||
XM_011519186.1 | XP_011517488.1 | 4 | 495 | |||
XM_011519187.2 | XP_011517489.1 | 3 | 535 | |||
XM_017015286.1 | XP_016870775.1 | 5 | 815 | |||
XM_017015287.1 | XP_016870776.1 | 4 | 495 | |||
GenBank mRNA | AK291603.1 | - | 4 | 437 | ||
AK291809.1 | - | 4 | 476 | |||
AK292790.1 | - | 4 | 476 | |||
AK298046.1 | - | 4 | 466 | |||
AK300718.1 | - | 3 | 294 | |||
AK310974.1 | - | 3 | 535 | |||
AK316266.1 | - | 5 | 764 | |||
BC013049.1 | - | 3 | 408 | 3882448 | ||
BC053604.1 | - | 3 | 409 | 6008684 | ||
BF982227.1 | - | 482 |