Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPS8L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319952.1 | NP_001306881.1 | 3 | 337 | ||
NM_024526.3 | NP_078802.2 | 4 | 392 | |||
NM_133181.3 | NP_573444.2 | 4 | 392 | |||
NM_139053.2 | NP_620641.1 | 4 | 392 | |||
XM_011542132.1 | XP_011540434.1 | 4 | 290 | |||
XM_011542133.1 | XP_011540435.1 | 4 | 290 | |||
XM_011542134.2 | XP_011540436.1 | 3 | 289 | |||
XM_011542135.2 | XP_011540437.1 | 4 | 407 | |||
XM_017002327.1 | XP_016857816.1 | 4 | 290 | |||
XM_017002328.1 | XP_016857817.1 | 4 | 290 | |||
XM_017002329.1 | XP_016857818.1 | 4 | 290 | |||
GenBank mRNA | AK025175.1 | - | 4 | 294 | ||
AK225235.1 | - | 4 | 294 | |||
AK290261.1 | - | 4 | 319 | |||
AK292198.1 | - | 4 | 316 | |||
AK307932.1 | - | 3 | 235 | |||
AK310979.1 | - | 4 | 352 | |||
AY074930.1 | - | 4 | 278 | |||
BC012926.1 | - | 4 | 312 | 3853503 | ||
CB159338.1 | - | 4 | 397 | |||
DA927692.1 | - | 4 | 289 |