Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139159.4 | NP_631898.3 | 13 | 1726 | ||
XM_005259673.2 | XP_005259730.1 | 14 | 1835 | |||
XM_011528405.1 | XP_011526707.1 | 15 | 1887 | |||
XM_011528406.1 | XP_011526708.1 | 16 | 1997 | |||
XM_011528408.1 | XP_011526710.1 | 13 | 1672 | |||
XM_011528410.1 | XP_011526712.1 | 14 | 1887 | |||
XM_017027453.1 | XP_016882942.1 | 15 | 2477 | |||
XM_017027454.1 | XP_016882943.1 | 16 | 2587 | |||
GenBank mRNA | AF452102.2 | - | 13 | 1746 | ||
AF542510.2 | - | 13 | 1705 | |||
AK054656.1 | - | 2 | 266 | |||
AK122645.1 | - | 7 | 776 | |||
AK122654.1 | - | 13 | 1729 | |||
AK131100.1 | - | 10 | 1295 | |||
AK131499.1 | - | 12 | 1239 | |||
AY172660.1 | - | 10 | 1391 | |||
AY374518.1 | - | 10 | 1381 | |||
BC000970.1 | - | 1 | 109 | |||
BC037948.2 | - | 13 | 1663 | 4577856 | ||
CR627380.1 | - | 5 | 501 | |||
DQ417928.1 | - | 11 | 1468 |