Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139159.4 | NP_631898.3 | 22 | 2900 | ||
XM_005259673.2 | XP_005259730.1 | 23 | 3009 | |||
XM_011528405.1 | XP_011526707.1 | 24 | 3061 | |||
XM_011528406.1 | XP_011526708.1 | 25 | 3171 | |||
XM_011528408.1 | XP_011526710.1 | 22 | 2846 | |||
XM_011528410.1 | XP_011526712.1 | 23 | 3061 | |||
XM_017027453.1 | XP_016882942.1 | 24 | 3651 | |||
XM_017027454.1 | XP_016882943.1 | 25 | 3761 | |||
GenBank mRNA | AF452102.2 | - | 22 | 2920 | ||
AK054656.1 | - | 11 | 1440 | |||
AK075030.1 | - | 4 | 501 | |||
AK122645.1 | - | 16 | 1950 | |||
AK122654.1 | - | 22 | 2903 | |||
AK131100.1 | - | 19 | 3420 | |||
AK131499.1 | - | 21 | 2413 | |||
AY172660.1 | - | 19 | 2565 | |||
BC000970.1 | - | 10 | 1283 | |||
BC037948.2 | - | 22 | 2837 | 4577856 | ||
CR627380.1 | - | 14 | 1675 | |||
DQ417928.1 | - | 20 | 2642 |