Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202498.1 | NP_001189427.1 | 7 | 1129 | ||
NM_003344.3 | NP_003335.1 | 7 | 1525 | |||
NM_182697.2 | NP_874356.1 | 5 | 1432 | |||
GenBank mRNA | AK091088.1 | - | 1 | 1473 | ||
AL832212.1 | - | 1 | 1534 | |||
Z29330.1 | - | 7 | 1173 | |||
Z29331.1 | - | 2 | 790 |