Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHURC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204063.1 | NP_001190992.1 | 4 | 2168 | ||
NM_001204064.1 | NP_001190993.1 | 3 | 2097 | |||
NM_145165.3 | NP_660148.3 | 4 | 2171 | |||
GenBank mRNA | AK057626.1 | - | 4 | 2078 | ||
BC020550.1 | - | 4 | 2118 | 4155545 |