Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080554.2 | NP_001074023.1 | 7 | 1370 | ||
NM_001080555.2 | NP_001074024.1 | 7 | 1246 | |||
NM_001206842.1 | NP_001193771.1 | 6 | 1177 | |||
NM_001206843.1 | NP_001193772.1 | 6 | 1069 | |||
NM_001206845.1 | NP_001193774.1 | 5 | 1000 | |||
NM_031289.3 | NP_112579.2 | 6 | 1277 | |||
NM_153823.3 | NP_722545.2 | 6 | 1153 | |||
XM_005253493.2 | XP_005253550.1 | 7 | 1300 | |||
XM_005253495.1 | XP_005253552.1 | 7 | 1424 | |||
XM_011520858.1 | XP_011519160.1 | 7 | 1327 | |||
XM_011520859.1 | XP_011519161.1 | 7 | 1244 | |||
XM_011520860.1 | XP_011519162.1 | 7 | 1451 | |||
XM_011520861.1 | XP_011519163.1 | 6 | 1228 | |||
XM_011520862.1 | XP_011519164.1 | 7 | 1368 | |||
XM_011520863.1 | XP_011519165.1 | 6 | 1352 | |||
GenBank mRNA | AB593019.1 | - | 6 | 1149 | ||
AB593020.1 | - | 6 | 1025 | |||
AK027894.1 | - | 6 | 1246 | |||
AK058045.1 | - | 8 | 1239 | |||
AK075288.1 | - | 7 | 1074 | |||
AK075311.1 | - | 5 | 842 | |||
AK075322.1 | - | 6 | 1023 | |||
AY358513.1 | - | 6 | 1131 | |||
BC001796.1 | - | 6 | 1223 | 3353821 | ||
BC033854.1 | - | 6 | 1120 | 5164943 | ||
BC112896.1 | - | 6 | 937 | 6145063 |