Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRPM3 SIRNA Assays ›
Gene Symbol:
TRPM3
Gene Name:
transient receptor potential cation channel subfamily M member 3
Gene Aliases:
GON-2, LTRPC3, MLSN2
Chromosome Location:
Chr.9: 70529063 - 71446989 on Build GRCh38
UniGene ID:
Hs.47288
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
131900
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRPM3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001007470.1
NP_001007471.1
8
997
NM_001007471.2
NP_001007472.2
7
1270
NM_020952.4
NP_066003.3
7
922
NM_024971.5
NP_079247.5
7
922
NM_206944.3
NP_996827.3
7
922
NM_206945.3
NP_996828.3
7
922
NM_206946.3
NP_996829.3
8
997
NM_206947.3
NP_996830.3
8
997
NM_206948.2
NP_996831.1
7
922
XM_011519035.2
XP_011517337.1
8
1222
XM_011519036.2
XP_011517338.1
8
4147
XM_011519037.2
XP_011517339.1
8
4133
XM_011519038.2
XP_011517340.1
8
4130
XM_011519039.2
XP_011517341.1
8
4131
XM_011519040.2
XP_011517342.1
7
1153
XM_011519041.2
XP_011517343.1
7
4055
XM_011519042.2
XP_011517344.1
7
1153
XM_011519043.2
XP_011517345.1
7
1186
XM_011519044.2
XP_011517346.1
7
4056
XM_011519045.2
XP_011517347.1
7
1147
XM_011519046.2
XP_011517348.1
7
2850
XM_011519047.2
XP_011517349.1
7
2820
XM_017015141.1
XP_016870630.1
8
1222
XM_017015142.1
XP_016870631.1
8
1222
XM_017015143.1
XP_016870632.1
8
1222
XM_017015144.1
XP_016870633.1
8
988
XM_017015145.1
XP_016870634.1
8
1666
XM_017015146.1
XP_016870635.1
8
1064
XM_017015147.1
XP_016870636.1
8
3099
XM_017015148.1
XP_016870637.1
8
1222
XM_017015149.1
XP_016870638.1
8
1222
XM_017015150.1
XP_016870639.1
8
1222
XM_017015151.1
XP_016870640.1
7
1165
XM_017015152.1
XP_016870641.1
8
2900
XM_017015153.1
XP_016870642.1
7
1157
XM_017015154.1
XP_016870643.1
8
1368
XM_017015155.1
XP_016870644.1
7
2822
XM_017015156.1
XP_016870645.1
3
345
XM_017015157.1
XP_016870646.1
7
2820
XM_017015158.1
XP_016870647.1
7
2818
XM_017015159.1
XP_016870648.1
7
913
XM_017015160.1
XP_016870649.1
7
1003
XM_017015161.1
XP_016870650.1
8
1222
GenBank mRNA
AB099661.1
-
4
567
AB099662.1
-
5
642
AB099663.1
-
4
567
AB099664.1
-
5
642
AB099665.1
-
4
567
AF325212.1
-
7
893
AF536748.1
-
4
567
AF536749.1
-
4
567
AF536750.1
-
4
567
AF536751.1
-
4
567
AF536752.1
-
4
567
AF536753.1
-
5
642
AJ505025.1
-
7
1270
AJ505026.1
-
7
1270
AK225732.1
-
7
913
AK308682.1
-
7
1098
BC022454.2
-
7
927
BC067733.1
-
8
959
BC094699.1
-
7
916
BC121821.1
-
5
768
BC134414.1
-
5
771
BC142972.1
-
7
939
KF987075.1
-
7
978
Back To Top
More Information
Panther Classification:
Molecular Function -
ion channel
transporter
Gene Ontology Categories:
Function(s)
Process(es)
cation transport
detection of temperature stimulus
sensory perception of temperature stimulus
protein tetramerization
calcium ion transmembrane transport
calcium activated cation channel activity
cation channel activity
calcium channel activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0