Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDGFC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016205.2 | NP_057289.1 | 1 | 574 | ||
NR_036641.1 | 1 | 574 | ||||
XM_011532124.2 | XP_011530426.1 | 1 | 18 | |||
XM_017008456.1 | XP_016863945.1 | 1 | 536 | |||
GenBank mRNA | AB033831.1 | - | 1 | 409 | ||
AF091434.1 | - | 1 | 574 | |||
AF244813.1 | - | 1 | 271 | |||
AF260738.1 | - | 1 | 245 | |||
AK300480.1 | - | 1 | 256 | |||
AK313817.1 | - | 1 | 304 | |||
AM922296.1 | - | 1 | 394 | |||
AY358493.1 | - | 1 | 367 | |||
BC094746.1 | - | 1 | 176 | |||
BC136662.1 | - | 1 | 495 | 9020652 |