Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDGFC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016205.2 | NP_057289.1 | 5, 6 | 1420 | ||
NR_036641.1 | 6, 7 | 1481 | ||||
XM_011532124.2 | XP_011530426.1 | 7, 8 | 1100 | |||
XM_017008455.1 | XP_016863944.1 | 5, 6 | 1049 | |||
XM_017008456.1 | XP_016863945.1 | 5, 6 | 1193 | |||
GenBank mRNA | AB033831.1 | - | 5, 6 | 1255 | ||
AF091434.1 | - | 5, 6 | 1420 | |||
AF244813.1 | - | 5, 6 | 1117 | |||
AF260738.1 | - | 5, 6 | 1091 | |||
AK300480.1 | - | 4, 5 | 906 | |||
AK304638.1 | - | 7, 8 | 763 | |||
AK313817.1 | - | 5, 6 | 1150 | |||
AM922296.1 | - | 6, 7 | 1301 | |||
AY358493.1 | - | 5, 6 | 1213 | |||
BC094746.1 | - | 5, 6 | 1021 | |||
BC136662.1 | - | 5, 6 | 1341 | 9020652 |