Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KBTBD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318716.1 | NP_001305645.1 | 4 | 1399 | ||
NM_001318717.1 | NP_001305646.1 | 4 | 1320 | |||
NM_001318718.1 | NP_001305647.1 | 4 | 1300 | |||
NM_001318719.1 | NP_001305648.1 | 4 | 1278 | |||
NM_001318720.1 | NP_001305649.1 | 4 | 1272 | |||
NM_001318721.1 | NP_001305650.1 | 4 | 1247 | |||
NM_001318722.1 | NP_001305651.1 | 4 | 1268 | |||
NM_001318723.1 | NP_001305652.1 | 4 | 1352 | |||
NM_001318724.1 | NP_001305653.1 | 4 | 1352 | |||
NM_001318725.1 | NP_001305654.1 | 4 | 1255 | |||
NM_016506.6 | NP_057590.3 | 4 | 1356 | |||
NM_018095.5 | NP_060565.4 | 4 | 1251 | |||
XM_017018009.1 | XP_016873498.1 | 4 | 1235 | |||
GenBank mRNA | AK001312.1 | - | 4 | 1205 | ||
AK001749.1 | - | 4 | 1292 | |||
AK091607.1 | - | 3 | 1929 | |||
AL713735.1 | - | 3 | 1181 | |||
BC002736.2 | - | 4 | 1293 | 3628290 | ||
CR457270.1 | - | 3 | 1139 |