Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316936.1 | NP_001303865.1 | 2 | 1629 | ||
NM_001316937.1 | NP_001303866.1 | 3 | 1702 | |||
NM_001316938.1 | NP_001303867.1 | 4 | 1772 | |||
NM_001316939.1 | NP_001303868.1 | 4 | 1746 | |||
NM_001316940.1 | NP_001303869.1 | 3 | 1624 | |||
NM_001316941.1 | NP_001303870.1 | 2 | 1655 | |||
NM_001316942.1 | NP_001303871.1 | 4 | 1800 | |||
NM_017703.2 | NP_060173.1 | 3 | 1728 | |||
XM_006722782.3 | XP_006722845.2 | 2 | 1479 | |||
XM_017026912.1 | XP_016882401.1 | 3 | 1604 | |||
GenBank mRNA | AK000195.1 | - | 3 | 1680 | ||
AK093760.1 | - | 3 | 1675 | |||
BC001586.2 | - | 3 | 1632 | 2966439 | ||
BX647073.1 | - | 2 | 1565 |