Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ST7L SIRNA Assays ›
Gene Symbol:
ST7L
Gene Name:
suppression of tumorigenicity 7 like
Gene Aliases:
FAM4B, ST7R, STLR
Chromosome Location:
Chr.1: 112517444 - 112620838 on Build GRCh38
UniGene ID:
Hs.201921
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
132059
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ST7L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308264.1
NP_001295193.1
5
777
NM_017744.4
NP_060214.2
6
986
NM_138727.3
NP_620055.1
6
935
NM_138728.2
NP_620056.1
6
986
NM_138729.3
NP_620057.1
6
986
XM_005270963.4
XP_005271020.1
6
509
XM_005270964.3
XP_005271021.1
5
601
XM_011541627.2
XP_011539929.1
6
695
XM_011541628.2
XP_011539930.1
6
695
XM_011541629.2
XP_011539931.1
6
709
XM_011541630.2
XP_011539932.1
6
1405
XM_011541631.2
XP_011539933.1
6
533
XM_017001534.1
XP_016857023.1
6
695
XM_017001535.1
XP_016857024.1
6
695
XM_017001536.1
XP_016857025.1
5
601
XM_017001537.1
XP_016857026.1
4
421
XM_017001538.1
XP_016857027.1
5
476
GenBank mRNA
AB081317.1
-
6
757
AB081318.1
-
6
706
AB081319.1
-
6
757
AB081320.1
-
6
757
AK021719.1
-
5
596
AK095837.1
-
4
294
AK097837.1
-
5
566
AK291042.1
-
6
687
AK301869.1
-
6
687
AK304004.1
-
6
644
BC027898.1
-
5
588
BC037830.1
-
6
1399
4812133
BC047392.1
-
6
712
5273549
BC065501.1
-
6
893
4417786
BX537988.1
-
7
848
BX640667.1
-
6
662
Back To Top
More Information
Gene Ontology Categories:
Function(s)
negative regulation of cell growth
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0