Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UHRF1BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017754.3 | NP_060224.3 | 14 | 2978 | ||
GenBank mRNA | AB126777.1 | - | 14 | 2813 | ||
AK122918.1 | - | 1 | 801 | |||
BC137120.1 | - | 14 | 2808 | 9021117 | ||
BC137127.1 | - | 14 | 2808 | 9021124 | ||
BX640641.1 | - | 14 | 2848 |