Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G2E3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308097.1 | NP_001295026.1 | 6 | 656 | ||
NM_017769.4 | NP_060239.2 | 7 | 697 | |||
XM_011536925.2 | XP_011535227.1 | 7 | 663 | |||
XM_011536926.1 | XP_011535228.1 | 7 | 697 | |||
GenBank mRNA | AB037754.1 | - | 7 | 649 | ||
AK093536.1 | - | 7 | 1684 | |||
AK295571.1 | - | 6 | 621 | |||
BC000973.2 | - | 7 | 643 | 3450488 |