Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLVCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195283.1 | NP_001182212.1 | 10 | 1502 | ||
NM_017791.2 | NP_060261.2 | 10 | 2384 | |||
GenBank mRNA | AK000378.1 | - | 10 | 2332 | ||
AK027804.1 | - | 9 | 1272 | |||
AY260572.1 | - | 10 | 2364 | |||
AY260577.1 | - | 10 | 2364 | |||
BC019087.2 | - | 10 | 2347 | 4866427 |