Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSGEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017807.3 | NP_060277.1 | 2 | 580 | ||
GenBank mRNA | AB050442.1 | - | 2 | 264 | ||
AF052137.1 | - | 2 | 243 | |||
AJ271669.1 | - | 2 | 155 | |||
AK000418.1 | - | 2 | 284 | |||
BC032310.1 | - | 2 | 313 | 5211591 | ||
BX428958.2 | - | 2 | 558 | |||
CR457232.1 | - | 2 | 155 |