Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPNPEP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003399.5 | NP_003390.4 | 21 | 3114 | ||
GenBank mRNA | AF086417.1 | - | 1 | 336 | ||
BC126174.1 | - | 21 | 3031 | 8991890 | ||
BC143901.1 | - | 21 | 3031 | 9052419 | ||
U90724.2 | - | 21 | 3212 |