Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127649.2 | NP_001121121.1 | 5 | 3763 | ||
NM_001199319.1 | NP_001186248.1 | 5 | 3442 | |||
NM_017929.5 | NP_060399.1 | 6 | 3589 | |||
GenBank mRNA | AK000065.1 | - | 7 | 1863 | ||
AK000702.1 | - | 7 | 2148 | |||
BG257217.1 | - | 1 | 463 |