Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017948.5 | NP_060418.4 | 3, 4 | 532 | ||
NR_046106.1 | 3, 4 | 532 | ||||
XM_006717166.3 | XP_006717229.1 | 3, 4 | 398 | |||
XM_006717167.3 | XP_006717230.1 | 3, 4 | 520 | |||
XM_006717168.3 | XP_006717231.1 | 3, 4 | 291 | |||
XM_006717172.3 | XP_006717235.1 | 3, 4 | 303 | |||
XM_011518824.2 | XP_011517126.1 | 2, 3 | 116 | |||
XM_017014876.1 | XP_016870365.1 | 3, 4 | 398 | |||
XM_017014878.1 | XP_016870367.1 | 3, 4 | 429 | |||
XM_017014879.1 | XP_016870368.1 | 3, 4 | 261 | |||
XM_017014880.1 | XP_016870369.1 | 3, 4 | 303 | |||
GenBank mRNA | AB105104.1 | - | 3, 4 | 498 | ||
AB109030.1 | - | 2, 3 | 195 | |||
AK001049.1 | - | 3, 4 | 414 | |||
AK297355.1 | - | 3, 4 | 415 | |||
AK307408.1 | - | 3, 4 | 423 | |||
CR627363.1 | - | 3, 4 | 276 | |||
DA470513.1 | - | 3, 4 | 522 |