Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIM1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039775.3 | NP_001034864.2 | 20 | 5135 | ||
XM_005245918.2 | XP_005245975.1 | 20 | 5121 | |||
XM_011541672.1 | XP_011539974.1 | 19 | 5085 | |||
XM_011541673.2 | XP_011539975.1 | 20 | 5360 | |||
GenBank mRNA | AK000902.1 | - | 4 | 1742 | ||
AK095339.1 | - | 19 | 2179 | |||
BC136870.1 | - | 16 | 1877 | 9020861 |