Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCWPW1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258008.1 | NP_001244937.1 | 14 | 1678 | ||
NM_017984.4 | NP_060454.3 | 14 | 1675 | |||
XM_005250479.2 | XP_005250536.1 | 14 | 2112 | |||
XM_005250480.2 | XP_005250537.1 | 13 | 2031 | |||
XM_006716035.3 | XP_006716098.1 | 14 | 2112 | |||
XM_006716036.2 | XP_006716099.1 | 14 | 1616 | |||
XM_006716037.3 | XP_006716100.1 | 14 | 2109 | |||
XM_006716038.3 | XP_006716101.1 | 14 | 2109 | |||
XM_006716040.3 | XP_006716103.1 | 13 | 2031 | |||
XM_017012379.1 | XP_016867868.1 | 13 | 2028 | |||
XM_017012380.1 | XP_016867869.1 | 13 | 1532 | |||
GenBank mRNA | AK000919.1 | - | 9 | 1062 | ||
AK093038.1 | - | 14 | 1616 | |||
AK300745.1 | - | 14 | 1534 | |||
AK302108.1 | - | 13 | 1412 | |||
AL136735.1 | - | 14 | 1546 | |||
BC002725.1 | - | 11 | 1376 | 3631266 | ||
KU178677.1 | - | 9 | 938 | |||
KU178678.1 | - | 9 | 941 | |||
KU178679.1 | - | 9 | 938 | |||
KU178680.1 | - | 9 | 941 | |||
KU178681.1 | - | 9 | 938 | |||
KU178682.1 | - | 8 | 857 |