Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUFY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042417.1 | NP_001035882.1 | 9 | 860 | ||
NM_001278225.1 | NP_001265154.1 | 9 | 788 | |||
NM_017987.4 | NP_060457.4 | 10 | 1289 | |||
NR_103475.1 | 10 | 1137 | ||||
NR_103476.1 | 10 | 962 | ||||
XM_005269953.4 | XP_005270010.1 | 9 | 1187 | |||
XM_005269955.4 | XP_005270012.1 | 9 | 868 | |||
XM_005269956.4 | XP_005270013.1 | 10 | 1289 | |||
XM_005269957.4 | XP_005270014.1 | 9 | 857 | |||
XM_006717911.3 | XP_006717974.1 | 10 | 960 | |||
XM_011539942.2 | XP_011538244.1 | 10 | 1701 | |||
GenBank mRNA | AB040970.1 | - | 10 | 898 | ||
AF411980.1 | - | 9 | 809 | |||
AF461266.1 | - | 9 | 868 | |||
AK127019.1 | - | 10 | 1289 | |||
AK294211.1 | - | 9 | 771 | |||
AY528716.1 | - | 10 | 927 | |||
BC041092.1 | - | 9 | 831 | 5731891 |