Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006720187.2 | XP_006720250.1 | 28, 29 | 4531 | ||
XM_017021397.1 | XP_016876886.1 | 28, 29 | 4534 | |||
GenBank mRNA | AK128275.1 | - | 9, 10 | 979 | ||
AK302641.1 | - | 1, 2 | 155 | |||
BC015016.2 | - | 2, 3 | 262 |