Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MANSC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018050.2 | NP_060520.2 | 4 | 1263 | ||
XM_017019531.1 | XP_016875020.1 | 5 | 1467 | |||
GenBank mRNA | AK001160.1 | - | 5 | 1168 | ||
AK023622.1 | - | 4 | 1263 | |||
AK298681.1 | - | 2 | 898 | |||
AY358563.1 | - | 4 | 1230 | |||
BC032998.2 | - | 4 | 1280 | 4824116 |