Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018110.3 | NP_060580.2 | 9 | 1540 | ||
XM_005256042.3 | XP_005256099.1 | 7 | 1599 | |||
XM_005256043.2 | XP_005256100.1 | 8 | 1482 | |||
GenBank mRNA | AK001350.1 | - | 9 | 1467 | ||
BC001540.2 | - | 8 | 1239 | 2988350 | ||
BC003541.1 | - | 9 | 1482 | 3611472 |