Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018141.3 | NP_060611.2 | 4 | 298 | ||
XM_005249206.2 | XP_005249263.1 | 4 | 307 | |||
XM_011514724.2 | XP_011513026.1 | 5 | 302 | |||
XM_011514725.2 | XP_011513027.1 | 5 | 294 | |||
XM_017010996.1 | XP_016866485.1 | 4 | 299 | |||
GenBank mRNA | AF113220.1 | - | 4 | 258 | ||
AF277180.1 | - | 4 | 260 | |||
AK001429.1 | - | 4 | 267 | |||
AK098446.1 | - | 4 | 261 | |||
AK298219.1 | - | 4 | 281 | |||
AK316599.1 | - | 4 | 264 | |||
BC012560.1 | - | 4 | 268 | 4277803 |