Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC61A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142627.2 | NP_001136099.1 | 6 | 537 | ||
NM_001142628.1 | NP_001136100.1 | 5 | 471 | |||
NM_018144.3 | NP_060614.2 | 6 | 537 | |||
NR_024576.2 | 6 | 537 | ||||
NR_024577.2 | 7 | 670 | ||||
GenBank mRNA | AF346603.1 | - | 6 | 478 | ||
AK057532.1 | - | 7 | 611 | |||
AK292295.1 | - | 6 | 487 | |||
AK301841.1 | - | 5 | 424 | |||
BC026179.1 | - | 6 | 487 | 4823130 | ||
BP212860.1 | - | 6 | 537 |